Få mere at vide
Adiponitrile-d8, TRC
Artikelnummer. 30319324
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30319324 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319324 Leverandør Toronto Research Chemicals Leverandørnr. TRCA291602100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Adiponitrile-d8
CAS 34006-32-1
Molekylær formel C6 D8 N2
Mængde 100 mg
Synonym Hexanedinitrile-d8 (9CI), Adiponitrile-d8 (8CI), 2,2,3,3,4,4,5,5-Octadeuteriohexanedinitrile, Hexanedinitrile-D8, 1,4-Dicyanobutane-2,2,3,3,4,4,5,5-d8, Adipic dinitrile-d8, Adipodinitrile-d8, Butanedicarbonitrile-d8
InChI formel InChI=1S/C6H8N2/c7-5-3-1-2-4-6-8/h1-4H2/i1D2,2D2,3D2,4D2
SMIL [2H]C([2H])(C#N)C([2H])([2H])C([2H])([2H])C([2H])([2H])C#N
IUPAC navn 2,2,3,3,4,4,5,5-octadeuteriohexanedinitrile
Molekylvægt (g/mol) 116.19
Formel vægt 116.119