missing translation for 'onlineSavingsMsg'
Få mere at vide

Adipokinetic Hormone (Libellula Auripennis), TRC

Artikelnummer. 30327931
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327931 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30327931 Leverandør Toronto Research Chemicals Leverandørnr. TRCA29165025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Adipokinetic Hormone (Libellula Auripennis)
CAS 129536-34-1
Molekylær formel C46H61N11O12
Mængde 25 mg
Synonym 5-oxo-L-prolyl-L-valyl-L-asparaginyl-L-phenylalanyl-L-threonyl-L-prolyl-L-seryl-L-Tryptophanamide, Chromatophorotropin, red-pigment-concentrating (Pandalus borealis) 2-L-valine-5-L-threonine-7-L-serine-, Lia-AKH, Libau-AKH
InChI formel InChI=1S/C46H61N11O12/c1-23(2)37(55-40(63)29-15-16-36(61)50-29)45(68)53-32(20-35(47)60)41(64)52-31(18-25-10-5-4-6-11-25)42(65)56-38(24(3)59)46(69)57-17-9-14-34(57)44(67)54-33(22-58)43(66)51-30(39(48)62)19-26-21-49-28-13-8-7-12-27(26)28/h4-8,10-13,21,23-24,29-34,37-38,49,58-59H,9,14-20,22H2,1-3H3,(H2,47,60)(H2,48,62)(H,50,61)(H,51,66)(H,52,64)(H,53,68)(H,54,67)(H,55,63)(H,56,65)/t24-,29-,30?,31-,32+,33?,34?,37+,38+/m1/s1
SMIL CC(C)[C@H](NC(=O)[C@H]1CCC(=O)N1)C(=O)N[C@@H](CC(=O)N)C(=O)N[C@H](Cc2ccccc2)C(=O)N[C@@H]([C@@H](C)O)C(=O)N3CCCC3C(=O)NC(CO)C(=O)NC(Cc4c[nH]c5ccccc45)C(=O)N
IUPAC navn (2S)-N-[(2R)-1-[[(2S,3R)-1-[2-[[1-[[1-amino-3-(1H-indol-3-yl)-1-oxopropan-2-yl]amino]-3-hydroxy-1-oxopropan-2-yl]carbamoyl]pyrrolidin-1-yl]-3-hydroxy-1-oxobutan-2-yl]amino]-1-oxo-3-phenylpropan-2-yl]-2-[[(2S)-3-methyl-2-[[(2R)-5-oxopyrrolidine-2-carbonyl]amino]butanoyl]amino]butanediamide
Molekylvægt (g/mol) 960.04
Formel vægt 959.4501
Produkttitel
Vælg et problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.