Få mere at vide
Adipic Acid-d4 (Major), TRC
Artikelnummer. 30321256
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30321256 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321256 Leverandør Toronto Research Chemicals Leverandørnr. TRCA29159450MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Adipic Acid-d4
CAS 121311-78-2
Molekylær formel C6 D4 H6 O4
Mængde 50 mg
Synonym Hexanedioic-3,3,4,4-d4 acid (9CI), Hexanedioic acid-3,3,4,4-D4, Hexanedioic acid-d4, Adipic Acid-d4
InChI formel InChI=1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)/i1D2,2D2
SMIL [2H]C([2H])(CC(=O)O)C([2H])([2H])CC(=O)O
IUPAC navn 3,3,4,4-tetradeuteriohexanedioic acid
Molekylvægt (g/mol) 150.166
Formel vægt 150.083