Få mere at vide
Adipic Acid-13C6, TRC
Artikelnummer. 30006758
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006758 100 mg
30007855 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006758 Leverandør Toronto Research Chemicals Leverandørnr. TRCA291593100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 942037-55-0
Molekylær formel 13C6H10O4
Molekylvægt (g/mol) 152.1
Synonym 1,4-Butanedicarboxylic Acid-13C6, 1,6-Hexanedioic Acid-13C6, Acifloctin-13C6, Acinetten-13C6, Adilactetten-13C6, Adipinic Acid-13C6, Asapic-13C6, E 355-13C6, Inipol DS-13C6, NSC 7622-13C6, NSC 87836-13C6
IUPAC navn (1,2,3,4,5,6-13C6)hexanedioic acid
SMIL O[13C]([13C]([H])([H])[13C]([H])([H])[13C]([H])([H])[13C]([H])([H])[13C](O)=O)=O
Tekniske data
Kemisk navn eller materiale Adipic Acid-13C6
Mængde 100 mg
InChI formel InChI=1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)/i1+1,2+1,3+1,4+1,5+1,6+1