Få mere at vide
Adiphenine Hydrochloride, Mikromol™
Beskrivelse
Adiphenine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Adiphenine Hydrochloride |
| CAS | 50-42-0 |
| Analyt- eller komponentnavne | Adiphenine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H25 N O2 . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Adiphenine Hydrochloride, Diphenylacetic acid 2-(diethylamino)ethyl ester hydrochloride, 2-Diethylaminoethyl diphenylacetate hydrochloride, Adifenin hydrochloride, Adiphen hydrochloride, Adiphenine hydrochloride, Difacil hydrochloride, Diphacil hydrochloride, NSC 129224, Patrovina, Paxil, Paxil (anticholinergic), Sentiv, Spasmolytin, Spasmolytin hydrochloride, Spasnil, Trasentin hydrochloride, Trasentine hydrochloride, Vegantin, alpha-Phenylbenzeneacetic acid 2-(diethylamino)ethyl ester hydrochloride, beta-Diethylaminoethyl diphenylacetate hydrochloride |
| InChI formel | InChI=1S/C20H25NO2.ClH/c1-3-21(4-2)15-16-23-20(22)19(17-11-7-5-8-12-17)18-13-9-6-10-14-18;/h5-14,19H,3-4,15-16H2,1-2H3;1H |
| SMIL | Cl.CCN(CC)CCOC(=O)C(c1ccccc1)c2ccccc2 |
| IUPAC navn | 2-(diethylamino)ethyl 2,2-diphenylacetate;hydrochloride |
| Vis mere |