Få mere at vide
Adiphenine hydrochloride, MedChemExpress
Artikelnummer. 16408276
Encompass_Preferred
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
16408276 100 mg
1 options
Artikelnummer. 16408276 Leverandør Medchem Express Leverandørnr. HYB0379A
MedChemExpress Adiphenine hydrochloride is a non-competitive inhibitor of nicotinic acetylcholine receptor (nAChR), with an IC50s of 1.9, 1.8, 3.7, and 6.3 μM for α1, α3β4, α4β2, and α4β4, respectively. Adiphenine hydrochloride has anticonvulsant effects.
Adiphenine hydrochloride is a non-competitive inhibitor of nicotinic acetylcholine receptor (nAChR), with an IC50s of 1.9, 1.8, 3.7, and 6.3 μM for α1, α3β4, α4β2, and α4β4, respectively. Adiphenine hydrochloride has anticonvulsant effects.
Tekniske data
Kemisk navn eller materiale Adiphenine hydrochloride
Fysisk form Solid
CAS 50-42-0
Farve White
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C20H26ClNO2
Til brug med (applikation) Neuroscience-Neuromodulation
Opløselighedsinformation DMSO : ≥ 100 mg/mL (287.46 mM) ∣H2O : ≥ 50 mg/mL (143.73 mM)
SMIL O=C(OCCN(CC)CC)C(C1=CC=CC=C1)C2=CC=CC=C2.Cl
Molekylvægt (g/mol) 347.88
Formel vægt 347.88
Procent renhed 99.77%