Learn More
Epredia™ Adhesion Excell Slides Product Code.: 11882833
Epredia™ Adhesion Excell Slides
Product Code. 11882833 Shop All Epredia Products
Product Code. 11882833
Brand: Epredia™ J5800AMNZ
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
SuperFrost Excell slides are especially useful in applications involving high pH Heat Induced Epitope Retrieval (HIER), epon & methyl methacrylate sections, Laser Capture Microdissection (LCM) and cell culture.
  • No time wasted on slide-coating proceduress
  • No money wasted on expensive adhesives
  • Reliable and uniform coating
  • Minimal sample loss
  • Reliable and safe adhesive action
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 75 mm
Width (Metric) 25 mm
Color White
Product Content Correction
Product Title
Epredia™ Adhesion Excell Slides >