Få mere at vide
3M™ Adflo™ Standard Lithium Ion Battery
Shop alle 3M produkter
:
Battery Type: Lithium ion
Beskrivelse
- Fully charges in 4.5 hours. Expected operating time is 7-9 hours with new particle filter, new fully-charged battery and a standard airflow
- Batteries may be stored on the charger, fully charged, ready for immediate use
- For use with the 3M™ Adflo™ Powered Air Respirator
Tekniske data
Tekniske data
| Batterilevetid | 7 to 9 hr. |
| Batteritype | Lithium-ion |
| Certificeringer/compliance | EN 12941 |
| Produkttype | Standard Battery |
| Inkluderer | Particle filter (gas filter available as spare part) |