Få mere at vide
3M™ Adflo™ Lightweight Breathing Tube
Artikelnummer. 15719637 Shop alle 3M produkter
:
Color: Black
Artikelnummer. 15719637
Brand: 3M™ 7000000189
3M™ Adflo™ Lightweight Breathing Tube without QRS (Quick Release Swivel) is designed for use with the Speedglas welding helmet series 9000 and 3M™ Protective Visors.
  • This breathing tube can also be used for the Speedglas welding helmet series 9100 and Versaflo headtops M-series, by adding the helmet breathing tube adapter (part no 53 35 05)
  • You can extend the service life of the breathing tube by using a tube cover or a heavy-duty rubber breathing tube