missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Adenylate Cyclase 8 Antibody (ADCY8/7574), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24266MFV610
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Adenylate Cyclase 8 Monoclonal antibody specifically detects Adenylate Cyclase 8 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Tekniske data
Adenylate Cyclase 8 | |
Monoclonal | |
mFluor Violet 610 SE | |
AC8, ADCY3, adenylate cyclase 8 (brain), adenylate cyclase type 8, Adenylate cyclase type VIII, Adenylyl cyclase 8, adenylyl cyclase-8, brain, ATP pyrophosphate-lyase 8, Ca(2+)/calmodulin-activated adenylyl cyclase, EC 4.6.1.1, HBAC1 | |
Recombinant fragment (around aa1-200) of human Adenylate Cyclase 8 protein (exact sequence is proprietary) | |
0.1 mL | |
Lipid and Metabolism | |
114 | |
Store at 4°C in the dark. | |
IgG2 κ |
Immunohistochemistry (Paraffin) | |
ADCY8/7574 | |
50 mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Adenylate Cyclase 8 Antibody (ADCY8/7574), mFluor Violet 610 SE, Novus Biologicals™ > 0.1 mL, mFluor Violet 610 SE
Ser du en mulighed for forbedring?Del en indholdskorrektion