missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 5'-monophosphate monohydrate, MedChemExpress

Product Code. 16482019
Encompass_Preferred
Change view
Click to view available options
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16482019 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482019 Supplier Medchem Express Supplier No. HYA0181A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Adenosine 5'-monophosphate monohydrate is an adenosine A1 receptor agonist. Adenosine 5'-monophosphate monohydrate has significant antiviral activity against HSV-1 and HSV-2.

Adenosine 5'-monophosphate monohydrate is an adenosine A1 receptor agonist. Adenosine 5'-monophosphate monohydrate has significant antiviral activity against HSV-1 and HSV-2.

Specifications

Kemisk navn eller materiale Adenosine 5'-monophosphate monohydrate
Fysisk form Solid
CAS 18422-05-4
Farve White
Mængde 1 g
Renhedsgrad noter Research
Molekylær formel C10H16N5O8P
Til brug med (applikation) Neuroscience-Neuromodulation
Synonym 5'-AMP monohydrate
Opløselighedsinformation H2O : 5 mg/mL (13.69 mM; Need ultrasonic and warming)
SMIL O=P(O)(OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3N)O1)O)O)O.O
Molekylvægt (g/mol) 365.24
Formel vægt 365.24
Procent renhed 98.0%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.