missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine-5'-diphosphate disodium salt dihydrate, Affymetrix/USBTM
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Specifications
Physical Form | Powder |
Quantity | 5 g |
Beilstein | 3645656 |
IUPAC Name | disodium;[[(2 R,3 S,4 R,5 R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate |
Formula Weight | 507.2 |
Packaging | Plastic bottle |
Chemical Name or Material | Adenosine-5 '-Diphosphate Disodium Salt Dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'