Få mere at vide
Adenosine-5',5' '-d2, TRC
Artikelnummer. 30323259
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30323259 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323259 Leverandør Toronto Research Chemicals Leverandørnr. TRCA2804091MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Adenosine-5',5''-d2
CAS 82741-17-1
Molekylær formel C10H11D2N5O4
Mængde 1 mg
Synonym 9-β-D-Ribofuranosyl-9H-purin-6-amine-5',5''-d2, 9-β-D-Ribofuranosyladenine-5',5''-d2, Adenine Riboside-2'-d, Adenocard-5',5''-d2, Adenocor-5',5''-d2, Adenogesic-5',5''-d2, Adenoscan-5',5''-d2, Adrekar-5',5''-d2, Boniton-5',5''-d2, D-Adenosine-5',5''-d2, 1-(6-Amino-9H-purin-9-yl)-1-deoxy-β-D-ribofuranose-5',5''-d2, Myocol-5',5''-d2, NSC 7652-5',5''-d2
InChI formel InChI=1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7-,10-/m1/s1/i1D2
SMIL OC([2H])([2H])[C@@H]1[C@@H](O)[C@@H](O)[C@H](N2C(N=CN=C3N)=C3N=C2)O1
IUPAC navn (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-[dideuterio(hydroxy)methyl]oxolane-3,4-diol
Molekylvægt (g/mol) 269.26