Få mere at vide
Adenosine-2-d1, TRC
Artikelnummer. 30318041
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318041 10 mg
30312251 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318041 Leverandør Toronto Research Chemicals Leverandørnr. TRCA28041110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Adenosine-2-D
CAS 109923-50-4
Molekylær formel C10 D H12 N5 O4
Mængde 10 mg
Synonym Adenosine-2-d (9CI, ACI), Adenosine-2-D, 9-β-D-Ribofuranosyl-9H-purin-2d-6-amine, 9-β-D-Ribofuranosyl-9H-purin-6-amine-2d, Adenosine-2-d1
InChI formel InChI=1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7-,10-/m1/s1/i2D
SMIL [2H]c1nc(N)c2ncn([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)c2n1
IUPAC navn (2R,3R,4S,5R)-2-(6-amino-2-deuteriopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molekylvægt (g/mol) 268.2475
Formel vægt 268.103