Få mere at vide
Adenosine 2’,3’-Cyclic Phosphate Triethylammonium Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Adenosine 2',3'-Cyclic Phosphate Triethylammonium Salt |
| CAS | 73647-07-1 |
| Molekylær formel | C16H27N6O6P |
| Mængde | 50 mg |
| Synonym | Adenosine Cyclic 2’,3’-(Hydrogen Phosphate) Triethylammonium Salt, 2’,3’-cAMP Triethylammonium Salt, Cyclic 2’,3’-AMP Triethylammonium Salt, 2’,3’-Cyclic Adenosine Monophosphate Triethylammonium Salt, Adenosine Cyclic 2’,3’-Monophosphate Triethylammonium Salt |
| InChI formel | InChI=1S/C10H12N5O6P.C6H15N/c11-8-5-9(13-2-12-8)15(3-14-5)10-7-6(4(1-16)19-10)20-22(17,18)21-7;1-4-7(5-2)6-3/h2-4,6-7,10,16H,1H2,(H,17,18)(H2,11,12,13);4-6H2,1-3H3/t4-,6-,7-,10-;/m1./s1 |
| SMIL | O=P1([O-])O[C@@H]2[C@H](O1)[C@@H](CO)O[C@H]2N3C(N=CN=C4N)=C4N=C3.CC[NH+](CC)CC |
| IUPAC navn | [(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2-hydroxy-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-6-yl]methanol;N,N-diethylethanamine |
| Molekylvægt (g/mol) | 430.4 |