Få mere at vide
Tocris Bioscience™ Adenosine
:
50mg
Beskrivelse
Adenosine is a neurotransmitter that acts as the preferred endogenous agonist at all adenosine receptor subtypes.
Tekniske data
Tekniske data
| Mængde | 50 mg |
| Produkttype | Adenosine |
| Renhed | >99% |
| Molekylvægt (g/mol) | 267.24 |
| Inhibitorer | Adenosine |
For Research Use Only