Få mere at vide
Adenine-2-d1, TRC
Artikelnummer. 30328081
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328081 10 mg
30325360 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328081 Leverandør Toronto Research Chemicals Leverandørnr. TRCA28048110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Adenine-2-D
CAS 109923-52-6
Molekylær formel C5 D H4 N5
Mængde 10 mg
Synonym 9H-Purin-2-d-6-amine (ACI), 1H-Purin-2-d-6-amine (9CI), Adenine-2-d1, Adenine-2-D
InChI formel InChI=1S/C5H5N5/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H3,6,7,8,9,10)/i2D
SMIL [2H]c1nc(N)c2[nH]cnc2n1
IUPAC navn 2-deuterio-7H-purin-6-amine
Molekylvægt (g/mol) 136.1329
Formel vægt 136.0608