Få mere at vide
Adefovir-d4 Diethyl Ester, TRC
Artikelnummer. 30327863
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30327863 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327863 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6289172.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Adefovir-d4 Diethyl Ester
CAS 1189929-36-9
Molekylær formel C12H16D4N5O4P
Mængde 2.5 mg
Synonym P-[[2-(6-Amino-9H-purin-9-yl)ethoxy-d4]methyl]phosphonic Acid Diethyl Ester, [[2-(6-Amino-9H-purin-9-yl)ethoxy-d4]methyl]phosphonic Acid Diethyl Ester
InChI formel InChI=1S/C12H20N5O4P/c1-3-20-22(18,21-4-2)9-19-6-5-17-8-16-10-11(13)14-7-15-12(10)17/h7-8H,3-6,9H2,1-2H3,(H2,13,14,15)/i5D2,6D2
SMIL [2H]C([2H])(OCP(=O)(OCC)OCC)C([2H])([2H])n1cnc2c(N)ncnc12
IUPAC navn 9-[1,1,2,2-tetradeuterio-2-(diethoxyphosphorylmethoxy)ethyl]purin-6-amine
Molekylvægt (g/mol) 333.32