Learn More
BRAND™ Silicone Adapter for Micro-Classic
Silicone construction. Includes suction tube.
Brand: BRAND™ 25931
91.02 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Spare adapter for Micro-Classic
- Silicone construction
- Includes suction tube.
Specifications
Silicone | |
Micro-Classic | |
Suction tube | |
Bag |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-07-31
Product Content Correction
Product Title
BRAND™ Silicone Adapter for Micro-Classic > For use with Micro-Classic
Spot an opportunity for improvement?Share a Content Correction