Learn More
Eppendorf™ Adapter S-4x Universal Rotor Large
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4xUniversal-Large rotor.
Brand: Eppendorf™ 5920751000
2540.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Suitable for S-4xUniversal-Large rotor buckets
- Accommodates selected array of tubes, bottles, or cards
Specifications
Round Bottom | |
17.5 mm | |
S-4xUniversal-Large | |
24 |
24 x 14 mL Round Bottom Tubes | |
123 mm | |
24 | |
2/Pk. |
Product Content Correction
Product Title
Eppendorf™ Adapter S-4x Universal Rotor Large > 24 x 14 mL round-bottom tubes
Spot an opportunity for improvement?Share a Content Correction