Learn More
Lenz Laborglasintrumente Adapter Duran DIN 12249 Cones
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100219
Additional Details : Weight : 0.02400kg
 View more versions of this product
Product Code. 10792512
  • 183.38 DKK / Pack of 10
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
  • Cones
  • Acc. to DIN 12249 form B
  • Standard, conical ground cones
  • Made of DURAN™ tubing
Specifications
Specifications
NS 19/26
1.8 mm
17 mm
Duran Glass