Learn More
Lenz Laborglasintrumente Adapter Duran DIN 12249 Cones
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01100245
Additional Details : Weight : 0.40000kg
Description
- Cones
- Acc. to DIN 12249 form B
- Standard, conical ground cones
- Made of DURAN™ tubing
Specifications
NS 45/40 | |
2.3 mm | |
40 mm | |
Duran Glass |