Learn More
RETSCH Adapter Cable
Brand:  RETSCH 05.105.0301
26353.00 DKK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.99000kg
Product Code. 10516289
  • 27740.00 DKK / Each
Estimated Shipment: 05-08-2024
to see stock.
Specifications
Adapter Cable
2 m