Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand: HUBER 6431
Additional Details : Weight : 0.13000kg
Description
- Adapter
Spécification
Thread Size: M16 x 1 female to M30 x 1.5 male | |
Thermostat | |
Adapter |