Få mere at vide
Adapalene-d6 Methyl Ester, TRC
Artikelnummer. 30321043
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30321043 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321043 Leverandør Toronto Research Chemicals Leverandørnr. TRCA22501210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Adapalene-d6 Methyl Ester
CAS 1330264-23-7
Molekylær formel C29H24D6O3
Mængde 10 mg
Synonym Methyl 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthalenecarboxylate-d6, 6-(4-Methoxy-3-tricyclo[3.3.1.13,7]dec-1-ylphenyl)-2-naphthalenecarboxylic Acid Methyl Ester-d6
InChI formel InChI=1S/C29H30O3/c1-31-27-8-7-24(14-26(27)29-15-18-9-19(16-29)11-20(10-18)17-29)22-3-4-23-13-25(28(30)32-2)6-5-21(23)12-22/h3-8,12-14,18-20H,9-11,15-17H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])OC1=CC=C(C2=CC=C(C=C(C(OC([2H])([2H])[2H])=O)C=C3)C3=C2)C=C1C4(C[C@H]5C6)C[C@H]6C[C@H](C5)C4
IUPAC navn trideuteriomethyl 6-[3-(1-adamantyl)-4-(trideuteriomethoxy)phenyl]naphthalene-2-carboxylate
Molekylvægt (g/mol) 432.58