Få mere at vide
Adamantanine, TRC
Artikelnummer. 30317354
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317354 25 mg
30309828 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30317354 Leverandør Toronto Research Chemicals Leverandørnr. TRCA20824525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Adamantanine
CAS 42381-05-5
Molekylær formel C11 H17 N O2
Mængde 25 mg
Synonym 2-Aminotricyclo[3.3.1.13,7]decane-2-carboxylic Acid, 2-Aminoadamantane-2-carboxylic Acid, NSC 145160
InChI formel InChI=1S/C11H17NO2/c12-11(10(13)14)8-2-6-1-7(4-8)5-9(11)3-6/h6-9H,1-5,12H2,(H,13,14)
SMIL NC1(C2CC3CC(CC1C3)C2)C(=O)O
IUPAC navn 2-aminoadamantane-2-carboxylic acid
Molekylvægt (g/mol) 195.258
Formel vægt 195.126