Få mere at vide
Adamantane-d15-carboxylic Acid, TRC
Artikelnummer. 30179650
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30179650 2.5 mg 2.5mg
30179452 5 mg -
30179601 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179650 Leverandør Toronto Research Chemicals Leverandørnr. TRCA2082012.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 33830-12-5
Molekylær formel C11 D15 H O2
Molekylvægt (g/mol) 195.336
Synonym Tricyclo[3.3.1.13,7]decane-2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-d15-1-carboxylic acid (ACI), 1-Adamantane-d15-carboxylic acid (8CI), 1-Adamantanecarboxylic acid-d15, 1-Adamantyl-d15-carboxylic acid, 1-Carboxyadamantane-d15, Adamantoic acid-d15, Adamantoic Acid-D15
IUPAC navn 2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-pentadecadeuterioadamantane-1-carboxylic acid
SMIL [2H]C1([2H])C2([2H])C([2H])([2H])C3([2H])C([2H])([2H])C1([2H])C([2H])([2H])C(C(=O)O)(C3([2H])[2H])C2([2H])[2H]
Tekniske data
Kemisk navn eller materiale Adamantane-d15-carboxylic Acid
Mængde 2.5 mg
InChI formel InChI=1S/C11H16O2/c12-10(13)11-4-7-1-8(5-11)3-9(2-7)6-11/h7-9H,1-6H2,(H,12,13)/i1D2,2D2,3D2,4D2,5D2,6D2,7D,8D,9D
Formel vægt 195.2092