missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals ADA, 98+%
Click to view available options
Quantity:
25 g
100 g
500 g
Description
ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | ∼218°C (decomposition) |
Quantity | 25 g |
Beilstein | 1787181 |
Merck Index | 14,147 |
Solubility Information | Insoluble in water. |
Formula Weight | 190.16 |
Percent Purity | ≥98% |
Chemical Name or Material | ADA |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'