Få mere at vide
Acyclovir N-Methylene Dimer, TRC
Artikelnummer. 30133195
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133195 1 mg
30132882 5 mg
30010395 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133195 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1924501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1797131-64-6
Molekylær formel C17 H22 N10 O6
Molekylvægt (g/mol) 462.42
Synonym 2,2'-[Methylenediimino]bis[9-[(2-hydroxyethoxy)methyl]-1,9-dihydro-6H-purin-6-one], 9-(2-Hydroxyethoxymethyl)-2-[[[9-(2-hydroxyethoxymethyl)-6-oxo-3H-purin-2-yl]amino]methylamino]-3H-purin-6-one, Aciclovir N-Methylene Dimer, Acyclovir N-Methylene Dimer
IUPAC navn 9-(2-hydroxyethoxymethyl)-2-[[[9-(2-hydroxyethoxymethyl)-6-oxo-1H-purin-2-yl]amino]methylamino]-1H-purin-6-one
SMIL OCCOCn1cnc2C(=O)NC(=Nc12)NCNC3=Nc4c(ncn4COCCO)C(=O)N3
Tekniske data
Kemisk navn eller materiale Acyclovir N-Methylene Dimer
Analyt- eller komponentnavne Acyclovir N-Methylene Dimer
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C17H22N10O6/c28-1-3-32-8-26-6-20-10-12(26)22-16(24-14(10)30)18-5-19-17-23-13-11(15(31)25-17)21-7-27(13)9-33-4-2-29/h6-7,28-29H,1-5,8-9H2,(H2,18,22,24,30)(H2,19,23,25,31)
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 1 mg
Formel vægt 462.1724