Få mere at vide
Acyclovir-d4, TRC
Artikelnummer. 30323875
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30323875 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323875 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1924021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Aciclovir-d4
CAS 1185179-33-2
Renhedsgrad noter HPLC
Molekylær formel C8 D4 H7 N5 O3
Mængde 1 mg
Synonym Aciclovir-d4, Acyclovir-D4, 2-Amino-9-[(1,1,2,2-tetradeuterio-2-hydroxyethoxy)methyl]purin-6-ol
InChI formel InChI=1S/C8H11N5O3/c9-8-11-6-5(7(15)12-8)10-3-13(6)4-16-2-1-14/h3,14H,1-2,4H2,(H3,9,11,12,15)/i1D2,2D2
SMIL [2H]C([2H])(O)C([2H])([2H])OCn1cnc2C(=O)NC(=Nc12)N
IUPAC navn 2-amino-9-[(1,1,2,2-tetradeuterio-2-hydroxy-ethoxy)methyl]-1H-purin-6-one
Molekylvægt (g/mol) 229.229
Formel vægt 229.1113
Procent renhed >95