missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acycloguanosine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Inhibits cytomegalovirus replication; induces apoptosis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 256°C to 260°C |
Color | White |
Quantity | 1 g |
Merck Index | 14,146 |
Solubility Information | Soluble in 1M HCl at 50mg/ml; Soluble in water at 0;7mg/ml; Also soluble in DMSO |
Formula Weight | 225.2 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Acycloguanosine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'