missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTR3C Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06502AF700
Additional Details : Weight : 0.00970kg
Description
ACTR3C Polyclonal antibody specifically detects ACTR3C in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
ACTR3C | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
653857 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
Actin-Related Arp11, Actin-related protein 11, Arp11, ARP3 Actin-Related Protein 3 Homolog C (Yeast) | |
Produced in rabbits immunized with E. coli-derived Human ACTR3C fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |