Få mere at vide
Solstice Activator Reagent, 0.5M ETT in Acetonitrile
Artikelnummer. 17195451 Shop alle Honeywell Fluka produkter
Skift visning
Mængde:
4 x 2,5 l
Emballagetype:
GLASFLASKE, GL45
Pakningsstørrelse:
2.5 L
Artikelnummer. Mængde Emballagetype unitSize
17195451 4 x 2,5 l GLASFLASKE, GL45 2.5 L
1 options
Artikelnummer. 17195451 Leverandør Solstice Leverandørnr. BR7252.5GL
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
BioSyn™, for DNA/RNA synthesis
Solstice BioSyn™ reagents and solvents for DNA/RNA synthesis
  • Solstice BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Tekniske data
CAS 75-05-8
Tæthed 0.809 g/cm3
Emballagetype GLASFLASKE, GL45
Mængde 4 x 2,5 l