missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal, DARCO™, 4 to 12mesh, Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Description
- DARCO is a registered trademark of Norit Americas, Inc.
Specifications
Melting Point | 3550°C |
Color | Black |
pH | 6 to 9 |
Mesh Size | 4 to 12mesh |
Packaging | Poly Bottle |
Quantity | 1 kg |
UN Number | NONH for all modes of transport |
Formula Weight | 12.01g/mol |
Grade | DARCO™ |
Physical Form | Granules |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'