Learn More
Ansell ActivArmr Gloves Class 0 - RIG014R
ActivArmr Gloves Class 0 - RIG014R are durable electrical protection gloves, ensuring safety and everyday usability.
765.00 DKK - 7600.00 DKK
Specifications
Arc Flash | Arc Rating of APC 1, Arc flash certified as per IEC 614821-2:2014 and ATSM F2675:2021 standards |
---|---|
Certifications/Compliance (Europe) | CE Number, Ecovadis label, EN ISO 21420:2020, EN 60903:2003, PPE category ELECTRICAL, Category III, Arc Rating of APC 1, ASTM D120, EN388:2016, REACH Compliant |
Color | Red |
Hand | Specific |
Material | Natural Rubber |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17888128
|
Ansell
RIG014R070 |
7 |
7600.00 DKK
10 pairs |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||||
17898128
|
Ansell
RIG014R080 |
8 |
7600.00 DKK
10 pairs |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||||
17808138
|
Ansell
RIG014R090 |
9 |
7600.00 DKK
10 pairs |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||||
17868128
|
Ansell
RIG014R100 |
10 |
7600.00 DKK
10 pairs |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||||
17878128
|
Ansell
RIG014R110 |
11 |
7600.00 DKK
10 pairs |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||||
17975152
|
Ansell
RIG014R120 |
12 |
765.00 DKK
10 pairs |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||||
Description
- Electrical Insulating Gloves are made from natural rubber latex using a proprietary environmentally friendly dipping process for ultimate flexibility and dexterity.
- Their ergonomic hand-at-rest shape and non-splayed fingers reduce hand fatigue.
- The generous flared cuff allows room for clothing.
- The design also features a smooth-finish for easy donning and doffing.
- Certified for resistance to acid (Category A), Ozone (Category Z) and very low temperatures (Category C).
- Applications with risk of touch voltage AC maximum 1000 V (DC maximum 1500)
Specifications
Arc Rating of APC 1, Arc flash certified as per IEC 614821-2:2014 and ATSM F2675:2021 standards | |
Red | |
Natural Rubber | |
Applications with risk of touch voltage Electrical Contractors Hybrid engine manufacturing and repair Maintenance Repairs and Operations in all industrial environments (MRO) Mass Transit Repair and Maintenance Power maintenance, repair and transmission Telecom repair near electrical wires | |
14 in. | |
1000 VAC, 1500 DC |
CE Number, Ecovadis label, EN ISO 21420:2020, EN 60903:2003, PPE category ELECTRICAL, Category III, Arc Rating of APC 1, ASTM D120, EN388:2016, REACH Compliant | |
Specific | |
Electrical Insulating Gloves | |
Gauntlet | |
360 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell ActivArmr Gloves Class 0 - RIG014R