Learn More
Ansell ActivArmr Electrical Insulating Gloves Class 3, Black, 16 in.
ActivArmr Electrical Insulating Gloves provide the ultimate fit, comfort and performance for electrical workers' safety and protection.
1550.00 DKK
Specifications
Certifications/Compliance (Europe) CE Number, Category III, EN 420:2003 + A1:2009, EN 60903:2003, Class 3
Color Black
Hand Specific
Material Natural Latex Rubber
Product Type Electrical Protection Gloves
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17825611
View Documents
Ansell
RIG316B080
8
1550.00 DKK
1 pair
Only null left
Estimated Shipment: 11-08-2025
Add to basket
17835611
View Documents
Ansell
RIG316B090
9
1550.00 DKK
1 pair
Only null left
Estimated Shipment: 11-08-2025
Add to basket
17845611
View Documents
Ansell
RIG316B100
10
1550.00 DKK
1 pair
Only null left
Estimated Shipment: 11-08-2025
Add to basket
17855611
View Documents
Ansell
RIG316B110
11
1550.00 DKK
1 pair
Only null left
Estimated Shipment: 11-08-2025
Add to basket
Description
Description
  • Performance Profile comfort
  • Natural Rubber Latex for ultimate durability and flexibility
  • Ergonomically-designed hand at rest shape for less hand fatigue
  • Generous flared cuff allows room for clothing and improves ventilation
  • Case hardened finish provides smoother surface for easy donning and doffing performance and protection
  • Product meets all applicable NFPA, OSHA and CSA standards
  • Environmentally-Friendly
  • Proprietary dipping process
  • ISO 14001 certified manufacturing facility
Specifications
Specifications
CE Number, Category III, EN 420:2003 + A1:2009, EN 60903:2003, Class 3
Specific
Electrical Protection Gloves
Black
RIG316B
410 mm
Black
Natural Latex Rubber
Applications with risk of touch voltage, Electrical Contractors, Hybrid engine manufacturing and repair, Maintenance Repairs and Operations in all industrial environments (MRO), Mass Transit Repair and Maintenance, Power maintenance, repair and transmission, Telecom repair near electrical wires
Beaded/Rolled
16 in.
26500/39750
Documents
Product Certifications
Product Content Correction
Product Title
Ansell ActivArmr Electrical Insulating Gloves Class 3, Black, 16 in.