Learn More
Ansell ActivArmr Electrical Insulating Gloves Class 0, Black, 11 in.
ActivArmr Electrical Insulating Gloves provide the ultimate fit, comfort and performance for electrical workers' safety and protection.
735.00 DKK
Specifications
Certifications/Compliance (Europe) | CE Number, Category III, Class 0 |
---|---|
Color | Black |
Hand | Specific |
Material | Natural Latex Rubber |
Product Type | Electrical Insulating Gloves |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17875551
|
Ansell
RIG011B070 |
7 |
735.00 DKK
1 pair |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
17885551
|
Ansell
RIG011B080 |
8 |
735.00 DKK
1 pair |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
17895551
|
Ansell
RIG011B090 |
9 |
735.00 DKK
1 pair |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
17805561
|
Ansell
RIG011B100 |
10 |
735.00 DKK
1 pair |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
17815561
|
Ansell
RIG011B110 |
11 |
735.00 DKK
1 pair |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
17825561
|
Ansell
RIG011B120 |
12 |
735.00 DKK
1 pair |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
Description
- Performance Profile comfort
- Natural Rubber Latex for ultimate durability and flexibility
- Ergonomically-designed hand at rest shape for less hand fatigue
- Generous flared cuff allows room for clothing and improves ventilation
- Case hardened finish provides smoother surface for easy donning and doffing performance and protection
- Product meets all applicable NFPA, OSHA and CSA standards
- Environmentally-Friendly
- Proprietary dipping process
- ISO 14001 certified manufacturing facility
Specifications
CE Number, Category III, Class 0 | |
Specific | |
Electrical Insulating Gloves | |
Black | |
RIG011B | |
280 mm |
Black | |
Natural Latex Rubber | |
Applications with risk of touch voltage, Electrical Contractors, Hybrid engine manufacturing and repair, Maintenance Repairs and Operations in all industrial environments (MRO), Mass Transit Repair and Maintenance, Power maintenance, repair and transmission, Telecom repair near electrical wires | |
Beaded/Rolled | |
11 in. | |
1000/1500 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell ActivArmr Electrical Insulating Gloves Class 0, Black, 11 in.