Få mere at vide
Actinamine Dihydrochloride, TRC
Artikelnummer. 30138186
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138186 1 g
30006724 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138186 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1919501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 52049-70-4
Molekylær formel C8 H18 N2 O4 . 2 Cl H
Molekylvægt (g/mol) 279.16
Synonym myo-Inositol, 1,3-dideoxy-1,3-bis(methylamino)-, hydrochloride (1:2), myo-Inositol, 1,3-dideoxy-1,3-bis(methylamino)-, dihydrochloride (9CI), Actinamine dihydrochloride
IUPAC navn (1R,3S,4R,6S)-4,6-bis(methylamino)cyclohexane-1,2,3,5-tetrol;dihydrochloride
SMIL Cl.Cl.CN[C@@H]1[C@@H](O)[C@H](O)[C@@H](O)[C@H](NC)[C@@H]1O
Tekniske data
Kemisk navn eller materiale Actinamine Dihydrochloride
Analyt- eller komponentnavne Actinamine Dihydrochloride
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C8H18N2O4.2ClH/c1-9-3-5(11)4(10-2)7(13)8(14)6(3)12;;/h3-14H,1-2H3;2*1H/t3-,4+,5+,6+,7-,8-;;
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 1 g
Formel vægt 278.08
Procent renhed >95%