Få mere at vide
MedChemExpress ACTH (4-11)
Artikelnummer. 16444066
Encompass_Preferred
Skift visning
Mængde:
25 mg
Pakningsstørrelse:
25mg
Artikelnummer. Mængde unitSize
16444066 25 mg 25mg
1 options
Artikelnummer. 16444066 Leverandør MedChemExpress Leverandørnr. HYP1503
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
MedChemExpress ACTH (4-11), an adrenocorticotropin hormone fragment, possesses a weak α-melanocyte stimulating hormone (α-MSH) potency only at high doses (100 and 1000 nM).
Tekniske data
CAS 67224-41-3
Indhold og opbevaring Please store the product under the recommended conditions in the Certificate of Analysis.
Til brug med (applikation) Metabolism-protein/nucleotide metabolism
Molekylvægt (g/mol) 1090.26 g/mol
Produkttype Peptides
Mængde 25 mg
Forsendelsestilstand Room temperature
Synonym Adrenocorticotropic Hormone (4-11), human