Få mere at vide
ACT-709478, MedChemExpress
Artikelnummer. 16494688
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16494688 10 mg -
16480977 50 mg 50mg
16447407 5 mg -
16425526 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16494688 Leverandør Medchem Express Leverandørnr. HY112723
MedChemExpress ACT-709478 is a potent, selective, orally active, and brain penetrating T-type calcium channel blocker. ACT-709478 is used in the research of generalized epilepsies.
ACT-709478 is a potent, selective, orally active, and brain penetrating T-type calcium channel blocker. ACT-709478 is used in the research of generalized epilepsies.
Tekniske data
Kemisk navn eller materiale ACT-709478
CAS 1838651-58-3
Farve White
Renhedsgrad noter Research
Molekylær formel C22H18F3N5O
Mængde 10 mg
Opløselighedsinformation DMSO : ≥ 125 mg/mL (293.83 mM)
SMIL O=C(NC1=NN(CC2=NC=C(C#N)C=C2)C=C1)CC3=CC=C(C4(C(F)(F)F)CC4)C=C3
Molekylvægt (g/mol) 425.41
Formel vægt 425.41
Procent renhed 99.59%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation