missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
ACSM4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82549-0.1ML
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ACSM4 Polyclonal specifically detects ACSM4 in Human samples. It is validated for Western Blot.
Tekniske data
| ACSM4 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 341392 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| ACSM4 acyl-CoA synthetase medium-chain family member 4 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human ACSM4 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion