Få mere at vide
Acrylonitrile-d3, TRC
Artikelnummer. 30325753
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30325753 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325753 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1913755MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acrylonitrile (D3)
CAS 53807-26-4
Molekylær formel C3 D3 N
Mængde 5 mg
Synonym 2-Propenenitrile-2,3,3-d3 (9CI), Acrylonitrile-d3 (7CI), Perdeuterioacrylonitrile, Acrylonitrile-D3
InChI formel InChI=1S/C3H3N/c1-2-3-4/h2H,1H2/i1D2,2D
SMIL [2H]C(=C([2H])C#N)[2H]
IUPAC navn 2,3,3-trideuterioprop-2-enenitrile
Molekylvægt (g/mol) 56.0811
Formel vægt 56.0454