Få mere at vide
Acrylonitrile-2-d1, TRC
Artikelnummer. 30319500
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319500 5 mg
30311546 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319500 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1913835MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acrylonitrile-2-D
CAS 4635-82-9
Molekylær formel C3 D H2 N
Mængde 5 mg
Synonym 2-Propenenitrile-2-d (9CI), Acrylonitrile-2-d (6CI, 7CI, 8CI), 2-Deuterioprop-2-enenitrile, 2-Deutero-2-propenenitrile, α-Deuterated acrylonitrile, Acrylonitrile-2-D
InChI formel InChI=1S/C3H3N/c1-2-3-4/h2H,1H2/i2D
SMIL [2H]C(=C)C#N
IUPAC navn 2-deuterioprop-2-enenitrile
Molekylvægt (g/mol) 54.0688
Formel vægt 54.0328