Få mere at vide
Acrylic Acid-d3, TRC
Artikelnummer. 30327601
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30327601 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327601 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1913585MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acrylic Acid-d3
CAS 204259-63-2
Molekylær formel C3HD3O2
Mængde 5 mg
Synonym Acrylic-2,3,3-d3 Acid
InChI formel InChI=1S/C3H4O2/c1-2-3(4)5/h2H,1H2,(H,4,5)/i1D2,2D
SMIL OC(/C([2H])=C([2H])/[2H])=O
IUPAC navn 2,3,3-trideuterioprop-2-enoic acid
Molekylvægt (g/mol) 75.08