Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration
Optimized for a variety of applications requiring larger per well volume processing.
Brand:  Cytiva 8582
2142.25 DKK valid until 2025-08-31
Use promo code "24124" to get your promotional price.
 View more versions of this product
Product Code. 17104641
  • 2255.00 DKK / Pack of 10
Only null left
Estimated Shipment: 04-08-2025
Description
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTFE (Hydrophilic)
Non-sterile
10/Pk.
0.2 ÎĽm
Solvent Filtration
350 ÎĽL
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cytiva AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration > 350 ÎĽL; 0.2 ÎĽm; wwPTFE Membrane