Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Lysate Clearance
Optimized for a variety of applications requiring larger per well volume processing.
Brand: Cytiva (formerly Pall Lab) 8175
Additional Details : Weight : 0.68400kg
Description
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Borosilicate Glass w/o Binder, Supor (PES) | |
Non-sterile | |
5/Pk. |
3.0/0.2 μm | |
Polypropylene Housing | |
1 mL |