Få mere at vide
Acrolein Antibody (10A10), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59359AF700
Produktkode 18212724
  • 5690.00 DKK / 100µL
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Acrolein Monoclonal specifically detects Acrolein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
Acrolein
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
Protein G purified
Primary
Non-species specific
Purified
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
10A10
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Acrylic amide; Ethylene Carboxamide 2-Propenamide; 2-Propen-1-one; 2-propenal; Acraldehyde; Acrolein; Acrylic aldehyde
Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH).
100 μL
Specific for Acrolein modified proteins. Does not detect free acrolein. Does not cross-react with Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.
Store at 4C in the dark.
IgG1