Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Supor™ Membrane and Modified Acrylic Housing
These syringe filters are ideal for biological and general research.
Brand: Cytiva (formerly Pall Lab) 4506
Additional Details : Weight : 2.99250kg
Description
- Superior flow rates and higher throughputs than competitive devices
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
PES | |
Non-sterile | |
<70 μL | |
1000/Pk. | |
25 mm |
0.2 μm | |
2.8 cm2 | |
Modified Acrylic | |
55°C |