Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with Supor™ Membrane
These syringe filters are ideal for biological and general research.
1370.00 DKK - 24070.00 DKK
Specifications
Material (Membrane) | Glass Fiber, Supor (Hydrophilic PES) |
---|---|
Sterility | Non-sterile |
Filtration Area | 3.9 cm2 |
Hold up Volume (After Air Purge) | <125 μL |
Housing Material | Modified Acrylic |
Product Code | Brand | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
17164671
|
Cytiva (formerly Pall Lab)
AP-4426 |
0.45 μm | 1000/Pk. |
24070.00 DKK
Pack of 1000 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
17154671
|
Cytiva (formerly Pall Lab)
AP-4425 |
0.45 μm | 50/Pk. |
1425.00 DKK
Pack of 50 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
17124701
|
Cytiva (formerly Pall Lab)
AP-4799 |
0.2 μm | 50/Pk. |
1370.00 DKK
Pack of 50 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
Description
- Superior flow rates and higher throughputs than competitive devices
- For research applications only
Specifications
Glass Fiber, Supor (Hydrophilic PES) | |
3.9 cm2 | |
Modified Acrylic | |
25 mm |
Non-sterile | |
<125 μL | |
55°C |