Learn More
Cytiva (formerly Pall Lab) AcroCap™ Positive Pressure Devices
Designed for fast, positive-pressure filtration of aqueous solutions up to 3 L.
Brand: Cytiva (formerly Pall Lab) 4480
Additional Details : Weight : 0.41800kg
Description
- Sterilization by gamma irradiation
- Eliminates the risk of cytotoxic residuals from EtO sterilization
- Integral hydrophobic vent prevents air lock
- Product not approved for use in medical clinical surgical or other patient applications
Specifications
Hydrophilic PES | |
Sterile | |
1/4 in. hose barb inner taper | |
10/Pk. |
0.2 μm | |
15 cm | |
1/4 in. hose barb inner taper |